Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144612.6 | NP_653213.6 | 11 | 1462 | ||
XM_017025541.1 | XP_016881030.1 | 4 | 579 | |||
XM_017025542.1 | XP_016881031.1 | 4 | 579 | |||
XM_017025543.1 | XP_016881032.1 | 4 | 579 | |||
XM_017025544.1 | XP_016881033.1 | 4 | 579 | |||
XM_017025545.1 | XP_016881034.1 | 4 | 579 | |||
XM_017025546.1 | XP_016881035.1 | 4 | 579 | |||
XM_017025547.1 | XP_016881036.1 | 4 | 579 | |||
XM_017025548.1 | XP_016881037.1 | 11 | 1544 | |||
GenBank mRNA | AK127869.1 | - | 4 | 775 |