Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROKR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144773.3 | NP_658986.1 | 2 | 491 | ||
XM_011529159.2 | XP_011527461.1 | 2 | 1207 | |||
XM_017027646.1 | XP_016883135.1 | 2 | 491 | |||
GenBank mRNA | AB084081.1 | - | 1 | 244 | ||
AF506288.1 | - | 1 | 244 | |||
AK289995.1 | - | 2 | 491 | |||
BC104959.1 | - | 1 | 288 | 8143962 | ||
BC104961.1 | - | 1 | 288 | 8143964 | ||
EF577398.1 | - | 1 | 244 |
Set Membership: |
G-protein coupled receptor |