Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317037.1 | NP_001303966.1 | 14 | 2148 | ||
NM_144965.2 | NP_659402.1 | 14 | 2187 | |||
XM_005251733.1 | XP_005251790.1 | 13 | 1999 | |||
XM_005251734.1 | XP_005251791.1 | 13 | 2030 | |||
XM_006716970.1 | XP_006717033.1 | 14 | 2147 | |||
XM_006716971.1 | XP_006717034.1 | 13 | 1974 | |||
XM_011518283.1 | XP_011516585.1 | 13 | 2050 | |||
XM_011518284.1 | XP_011516586.1 | 13 | 2043 | |||
XM_011518285.1 | XP_011516587.1 | 12 | 1900 | |||
XM_011518288.1 | XP_011516590.1 | 11 | 1758 | |||
XM_011518289.2 | XP_011516591.1 | 14 | 2071 | |||
XM_011518290.1 | XP_011516592.1 | 13 | 2018 | |||
XM_011518291.1 | XP_011516593.1 | 13 | 2072 | |||
XM_011518292.1 | XP_011516594.1 | 12 | 1943 | |||
XM_011518293.1 | XP_011516595.1 | 14 | 2138 | |||
GenBank mRNA | AK057342.1 | - | 14 | 2172 | ||
AK074105.1 | - | 6 | 4504 | |||
AK128011.1 | - | 8 | 1742 | |||
AK302737.1 | - | 14 | 2133 | |||
BC031281.1 | - | 14 | 2185 | 5295890 |