Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146256.1 | NP_001139728.1 | 11 | 1934 | ||
NM_144969.2 | NP_659406.1 | 12 | 1961 | |||
GenBank mRNA | AK056374.1 | - | 12 | 1499 | ||
AK122885.1 | - | 11 | 1683 | |||
AK128587.1 | - | 11 | 1469 |