Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297743.1 | NP_001284672.1 | 2 | 1185 | ||
NM_145007.3 | NP_659444.2 | 5 | 2035 | |||
GenBank mRNA | AK090621.1 | - | 1 | 235 | ||
AK129784.1 | - | 1 | 262 | |||
AK310394.1 | - | 5 | 1712 | |||
AY095145.1 | - | 3 | 1423 | |||
AY154466.1 | - | 2 | 1324 | |||
AY168967.1 | - | 3 | 1412 | |||
AY168968.1 | - | 2 | 1084 | |||
AY226382.1 | - | 2 | 1324 | |||
BC034730.1 | - | 3 | 1412 | 4341197 |