Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195683.1 | NP_001182612.1 | 17 | 4318 | ||
NM_001195684.1 | NP_001182613.1 | 18 | 4154 | |||
NM_003243.4 | NP_003234.2 | 17 | 4321 | |||
NR_036634.1 | 18 | 4418 | ||||
GenBank mRNA | AJ251961.1 | - | 16 | 3868 | ||
BC099914.1 | - | 20 | 4311 | |||
HG764816.1 | - | 18 | 4154 | |||
L07594.1 | - | 18 | 4145 |