Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFT2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145169.2 | NP_660152.1 | 8 | 723 | ||
NR_130112.1 | 9 | 746 | ||||
NR_130113.1 | 7 | 651 | ||||
NR_130114.1 | 7 | 642 | ||||
GenBank mRNA | AK021489.1 | - | 2 | 1990 | ||
AK090795.1 | - | 1 | 1630 | |||
AL833217.1 | - | 2 | 3054 | |||
AW964442.1 | - | 7 | 573 | |||
BC018969.1 | - | 8 | 628 | 3959886 | ||
BC028388.1 | - | 8 | 2341 | |||
BI597156.1 | - | 9 | 709 |