Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145250.3 | NP_660293.1 | 3 | 803 | ||
GenBank mRNA | AK098687.1 | - | 2 | 660 | ||
AK292348.1 | - | 2 | 804 | |||
AY358794.1 | - | 3 | 1083 | |||
BC025410.1 | - | 3 | 730 | 4826406 | ||
BX161458.1 | - | 3 | 775 | |||
FJ237361.1 | - | 3 | 730 |