Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF511-PRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU937518.1 | - | 2 | 143 |
Gene Symbol: ZNF511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145806.3 | NP_665805.2 | 3 | 371 | ||
NR_130127.1 | 3 | 371 | ||||
GenBank mRNA | AK091711.1 | - | 3 | 319 | ||
AK292380.1 | - | 3 | 317 | |||
AL530248.3 | - | 3 | 366 | |||
BC015139.1 | - | 3 | 361 | |||
BC019897.1 | - | 3 | 341 | 5123355 | ||
HY159562.1 | - | 3 | 347 | |||
HY179580.1 | - | 3 | 370 |