Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145808.3 | NP_665807.1 | 4 | 1082 | ||
GenBank mRNA | AK056225.1 | - | 4 | 985 | ||
AK309816.1 | - | 2 | 782 | |||
AL834231.1 | - | 4 | 954 |
Gene Symbol: LUZP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128619.2 | NP_001122091.2 | 4 | 1082 |