Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147198.3 | NP_671731.1 | 5 | 535 | ||
GenBank mRNA | AY047610.1 | - | 5 | 535 |