Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024907.6 | NP_079183.4 | 6 | 1402 | ||
NM_148169.2 | NP_680474.1 | 6 | 1289 | |||
NR_104026.1 | 6 | 1440 | ||||
GenBank mRNA | AF386743.1 | - | 6 | 1329 | ||
AK021860.1 | - | 3 | 2142 | |||
AK057934.1 | - | 6 | 1289 | |||
BC012385.1 | - | 6 | 1241 | 3864760 |