Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 13 | 1985 | ||
NM_014491.3 | NP_055306.1 | 13 | 1988 | |||
NM_148898.3 | NP_683696.2 | 14 | 2063 | |||
NM_148900.3 | NP_683698.2 | 14 | 2039 | |||
NR_033766.1 | 16 | 1999 | ||||
NR_033767.1 | 14 | 2046 | ||||
XM_017012801.1 | XP_016868290.1 | 16 | 2669 | |||
GenBank mRNA | AF086040.1 | - | 3 | 378 | ||
AF337817.1 | - | 13 | 1764 | |||
AK131266.1 | - | 16 | 1983 | |||
AK296957.1 | - | 14 | 1919 | |||
AY144615.1 | - | 14 | 2058 | |||
BC126104.1 | - | 14 | 1720 | 8991820 | ||
BC143866.1 | - | 14 | 1720 | 9052382 | ||
BC143867.1 | - | 13 | 1659 | 9052383 | ||
CR749236.1 | - | 20 | 2369 | |||
DQ778626.1 | - | 8 | 1059 | |||
KJ534839.1 | - | 13 | 1689 | |||
KJ535079.1 | - | 12 | 1827 | |||
KJ535086.1 | - | 12 | 1613 |