Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042481.2 | NP_001035946.1 | 12 | 1525 | ||
NM_001267046.1 | NP_001253975.1 | 11 | 1325 | |||
NM_001267047.1 | NP_001253976.1 | 2 | 82 | |||
NM_152330.3 | NP_689543.1 | 11 | 1301 | |||
XM_006720029.1 | XP_006720092.1 | 11 | 1422 | |||
XM_006720030.1 | XP_006720093.1 | 11 | 1398 | |||
XM_011536423.1 | XP_011534725.1 | 12 | 1569 | |||
XM_011536424.1 | XP_011534726.1 | 13 | 1694 | |||
XM_017020967.1 | XP_016876456.1 | 11 | 1275 | |||
XM_017020968.1 | XP_016876457.1 | 11 | 1251 | |||
GenBank mRNA | AK055545.1 | - | 11 | 1236 | ||
AK126521.1 | - | 9 | 946 | |||
AK315764.1 | - | 11 | 1222 | |||
AL833158.1 | - | 12 | 1511 | |||
AL833380.1 | - | 6 | 540 | |||
AL833661.1 | - | 6 | 521 | |||
BC020521.2 | - | 11 | 1249 | 3914276 | ||
BC029870.1 | - | 2 | 82 | |||
BX161430.1 | - | 11 | 1301 | |||
BX647239.1 | - | 2 | 1870 | |||
BX647863.1 | - | 11 | 1213 | |||
BX648295.1 | - | 2 | 1871 |