Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003262.3 | NP_003253.1 | 3 | 238 | ||
XM_011513114.2 | XP_011511416.1 | 4 | 315 | |||
GenBank mRNA | BC003677.1 | - | 3 | 230 | ||
BC007040.1 | - | 3 | 230 | |||
BC012035.1 | - | 3 | 222 | 4706586 | ||
BC105971.1 | - | 321 | ||||
BX475495.1 | - | 3 | 239 | |||
BX648539.1 | - | 3 | 222 | |||
D87127.1 | - | 3 | 219 | |||
KC005990.1 | - | 3 | 207 | |||
U93239.1 | - | 3 | 219 |