Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080378.1 | NP_001073847.1 | 4 | 676 | ||
NM_001080379.1 | NP_001073848.1 | 3 | 718 | |||
NM_152410.2 | NP_689623.2 | 4 | 676 | |||
XM_005266825.4 | XP_005266882.1 | 3 | 801 | |||
XM_005266826.4 | XP_005266883.1 | 3 | 801 | |||
XM_006715344.3 | XP_006715407.1 | 3 | 801 | |||
XM_006715345.3 | XP_006715408.1 | 3 | 801 | |||
XM_011535461.2 | XP_011533763.1 | 3 | 801 | |||
XM_011535462.2 | XP_011533764.1 | 3 | 801 | |||
XM_011535466.1 | XP_011533768.1 | 3 | 801 | |||
XM_011535468.2 | XP_011533770.1 | 3 | 801 | |||
XM_011535469.2 | XP_011533771.1 | 3 | 801 | |||
XM_017010275.1 | XP_016865764.1 | 4 | 676 | |||
XM_017010276.1 | XP_016865765.1 | 4 | 667 | |||
XM_017010277.1 | XP_016865766.1 | 4 | 951 | |||
XM_017010278.1 | XP_016865767.1 | 4 | 601 | |||
XM_017010279.1 | XP_016865768.1 | 4 | 628 | |||
XM_017010280.1 | XP_016865769.1 | 3 | 801 | |||
XM_017010281.1 | XP_016865770.1 | 2 | 604 | |||
XM_017010282.1 | XP_016865771.1 | 3 | 520 | |||
GenBank mRNA | AF546872.1 | - | 3 | 718 | ||
AK057286.1 | - | 4 | 676 | |||
BC030642.2 | - | 3 | 688 | 4823539 | ||
BC044227.1 | - | 4 | 663 | 5187302 |