Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARNMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320497.1 | NP_001307426.1 | 3 | 359 | ||
NM_152420.2 | NP_689633.1 | 3 | 679 | |||
NR_135282.1 | 3 | 679 | ||||
XM_005251701.3 | XP_005251758.2 | 3 | 354 | |||
XM_011518223.2 | XP_011516525.1 | 3 | 705 | |||
XM_017014269.1 | XP_016869758.1 | 3 | 666 | |||
XM_017014270.1 | XP_016869759.1 | 4 | 612 | |||
XM_017014271.1 | XP_016869760.1 | 4 | 1104 | |||
GenBank mRNA | AK098661.1 | - | 2 | 106 | ||
AK299691.1 | - | 3 | 522 | |||
BC034033.1 | - | 3 | 643 | 4278547 | ||
BI560441.1 | - | 3 | 683 | |||
BX538061.1 | - | 2 | 196 | |||
BX647386.1 | - | 3 | 448 |