Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152467.3 | NP_689680.2 | 2 | 501 | ||
XM_011524706.2 | XP_011523008.1 | 4 | 634 | |||
GenBank mRNA | AK057224.1 | - | 2 | 464 | ||
AK301797.1 | - | 2 | 405 | |||
AK302141.1 | - | 2 | 449 | |||
AY495339.1 | - | 2 | 483 | |||
BC067753.1 | - | 2 | 488 | |||
BP371285.1 | - | 2 | 501 |