Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152551.3 | NP_689764.3 | 6, 7 | 784 | ||
XM_011514312.2 | XP_011512614.1 | 6, 7 | 720 | |||
GenBank mRNA | AK027220.1 | - | 1, 2 | 125 | ||
AK056796.1 | - | 6, 7 | 2423 | |||
AK292409.1 | - | 6, 7 | 762 | |||
AL832462.1 | - | 6, 7 | 785 | |||
BC062458.1 | - | 6, 7 | 746 | |||
BC114630.1 | - | 6, 7 | 752 | 40084896 | ||
BC114631.1 | - | 6, 7 | 753 | 40084903 |