Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278740.1 | NP_001265669.1 | 8 | 1965 | ||
NM_001278741.1 | NP_001265670.1 | 8 | 1735 | |||
NM_001278742.1 | NP_001265671.1 | 7 | 1627 | |||
NM_001278743.1 | NP_001265672.1 | 7 | 1651 | |||
NM_003270.3 | NP_003261.1 | 8 | 1673 | |||
XM_011531018.1 | XP_011529320.1 | 9 | 1475 | |||
GenBank mRNA | AB097016.1 | - | 8 | 1569 | ||
AB209876.1 | - | 8 | 1632 | |||
AF043906.1 | - | 8 | 1577 | |||
AF053453.1 | - | 8 | 1532 | |||
AK075072.1 | - | 8 | 1622 |