Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152573.3 | NP_689786.2 | 12 | 1889 | ||
XM_005251730.3 | XP_005251787.1 | 11 | 1869 | |||
XM_005251731.3 | XP_005251788.1 | 12 | 2380 | |||
GenBank mRNA | AK290986.1 | - | 12 | 1631 | ||
BC023566.1 | - | 12 | 1889 | 4138119 |