Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168339.1 | NP_001161811.1 | 10 | 988 | ||
NM_001168340.1 | NP_001161812.1 | 10 | 994 | |||
NM_001168341.1 | NP_001161813.1 | 8 | 787 | |||
NM_001168342.1 | NP_001161814.1 | 8 | 869 | |||
NM_152574.2 | NP_689787.2 | 10 | 994 | |||
XM_011517732.2 | XP_011516034.1 | 9 | 791 | |||
XM_017014310.1 | XP_016869799.1 | 12 | 1357 | |||
XM_017014311.1 | XP_016869800.1 | 11 | 1289 | |||
XM_017014312.1 | XP_016869801.1 | 9 | 859 | |||
GenBank mRNA | AK091187.1 | - | 10 | 880 | ||
AK093459.1 | - | 10 | 1061 | |||
AK298581.1 | - | 8 | 642 | |||
AK298998.1 | - | 10 | 966 | |||
AK301185.1 | - | 9 | 780 | |||
AK301427.1 | - | 8 | 785 | |||
AK310271.1 | - | 11 | 852 | |||
AK310482.1 | - | 10 | 994 | |||
BC042417.1 | - | 9 | 861 | |||
BC142985.1 | - | 10 | 967 | 8860428 |