Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168339.1 | NP_001161811.1 | 15 | 1485 | ||
NM_001168340.1 | NP_001161812.1 | 14 | 1452 | |||
NM_001168341.1 | NP_001161813.1 | 13 | 1284 | |||
NM_001168342.1 | NP_001161814.1 | 13 | 1366 | |||
NM_152574.2 | NP_689787.2 | 15 | 1491 | |||
XM_011517732.2 | XP_011516034.1 | 14 | 1288 | |||
XM_017014310.1 | XP_016869799.1 | 17 | 1854 | |||
XM_017014311.1 | XP_016869800.1 | 16 | 1786 | |||
XM_017014312.1 | XP_016869801.1 | 14 | 1356 | |||
GenBank mRNA | AK091187.1 | - | 15 | 1377 | ||
AK093459.1 | - | 15 | 1553 | |||
AK298581.1 | - | 13 | 1139 | |||
AK298998.1 | - | 14 | 1424 | |||
AK301427.1 | - | 13 | 1282 | |||
AK310482.1 | - | 15 | 1491 | |||
BC038592.1 | - | 2 | 201 | |||
BC042417.1 | - | 14 | 1358 | |||
BC142985.1 | - | 15 | 1464 | 8860428 |