Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152595.4 | NP_689808.2 | 1 | 1722 | ||
GenBank mRNA | AK057200.1 | - | 1 | 1723 | ||
AK094816.1 | - | 1 | 641 | |||
AK289491.1 | - | 1 | 1277 | |||
BC130441.1 | - | 1 | 1703 |