Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033214.2 | NP_149991.2 | 1 | 524 | ||
GenBank mRNA | AK292241.1 | - | 1 | 534 | ||
AK312768.1 | - | 1 | 516 | |||
BC029820.1 | - | 1 | 516 | 5169374 | ||
BC048274.1 | - | 1 | 523 | 5166566 | ||
BC058888.1 | - | 1 | 514 | 5172337 | ||
BI830891.1 | - | 1 | 521 | |||
X78712.1 | - | 1 | 524 |