Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152729.2 | NP_689942.2 | 2 | 232 | ||
XM_006715378.3 | XP_006715441.1 | 2 | 235 | |||
GenBank mRNA | AK290408.1 | - | 2 | 182 | ||
AK308337.1 | - | 2 | 215 | |||
BC108699.1 | - | 2 | 189 | 6061738 | ||
BC136832.1 | - | 2 | 191 | 9020823 | ||
BC136841.1 | - | 2 | 191 | 9020832 | ||
BM153683.1 | - | 2 | 238 | |||
BM808222.1 | - | 221 |