Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TBC1D32 SIRNA Assays ›
Gene Symbol:
TBC1D32
Gene Name:
TBC1 domain family member 32
Gene Aliases:
BROMI, C6orf170, C6orf171, bA301B7.2, bA57L9.1, dJ310J6.1
Chromosome Location:
Chr.6: 121079494 - 121335382 on Build GRCh38
UniGene ID:
Hs.121396
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
129543
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TBC1D32
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152730.5
NP_689943.4
12, 13
1450
NR_104452.1
12, 13
1468
XM_005266861.2
XP_005266918.1
12, 13
1450
XM_011535569.1
XP_011533871.1
12, 13
1450
XM_011535570.1
XP_011533872.1
13, 14
1498
XM_011535571.2
XP_011533873.1
13, 14
2151
XM_011535574.1
XP_011533876.1
12, 13
1446
XM_011535575.2
XP_011533877.1
13, 14
1498
XM_011535576.2
XP_011533878.1
13, 14
1498
XM_011535580.2
XP_011533882.1
3, 4
301
XM_011535582.2
XP_011533884.1
13, 14
1498
XM_011535585.2
XP_011533887.1
13, 14
1498
XM_017010397.1
XP_016865886.1
13, 14
1498
XM_017010398.1
XP_016865887.1
13, 14
2146
XM_017010399.1
XP_016865888.1
13, 14
1498
XM_017010400.1
XP_016865889.1
12, 13
1372
XM_017010401.1
XP_016865890.1
13, 14
1498
XM_017010402.1
XP_016865891.1
12, 13
1450
XM_017010403.1
XP_016865892.1
13, 14
1498
XM_017010405.1
XP_016865894.1
13, 14
1498
GenBank mRNA
AK091554.1
-
12, 13
1431
AK125385.1
-
1, 2
160
AK131446.1
-
12, 13
1448
Back To Top
More Information
Gene Ontology Categories:
Function(s)
lens development in camera-type eye
retinal pigment epithelium development
determination of left/right symmetry
heart development
embryonic digit morphogenesis
cilium morphogenesis
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
protein localization to cilium
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0