Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152730.5 | NP_689943.4 | 14 | 1656 | ||
NR_104452.1 | 14 | 1674 | ||||
XM_005266861.2 | XP_005266918.1 | 14 | 1656 | |||
XM_011535569.1 | XP_011533871.1 | 14 | 1656 | |||
XM_011535570.1 | XP_011533872.1 | 15 | 1704 | |||
XM_011535571.2 | XP_011533873.1 | 15 | 2357 | |||
XM_011535574.1 | XP_011533876.1 | 14 | 1652 | |||
XM_011535575.2 | XP_011533877.1 | 15 | 1704 | |||
XM_011535576.2 | XP_011533878.1 | 15 | 1704 | |||
XM_011535580.2 | XP_011533882.1 | 5 | 507 | |||
XM_011535582.2 | XP_011533884.1 | 15 | 1704 | |||
XM_011535585.2 | XP_011533887.1 | 15 | 1704 | |||
XM_017010397.1 | XP_016865886.1 | 15 | 1704 | |||
XM_017010398.1 | XP_016865887.1 | 15 | 2352 | |||
XM_017010399.1 | XP_016865888.1 | 15 | 1704 | |||
XM_017010400.1 | XP_016865889.1 | 14 | 1578 | |||
XM_017010401.1 | XP_016865890.1 | 15 | 1704 | |||
XM_017010402.1 | XP_016865891.1 | 14 | 1656 | |||
XM_017010403.1 | XP_016865892.1 | 15 | 1704 | |||
XM_017010405.1 | XP_016865894.1 | 15 | 1704 | |||
GenBank mRNA | AK091554.1 | - | 14 | 1637 | ||
AK125385.1 | - | 3 | 366 | |||
AK131446.1 | - | 14 | 1654 |