Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152743.3 | NP_689956.2 | 13 | 1964 | ||
XM_005249643.4 | XP_005249700.1 | 13 | 1926 | |||
XM_011515177.2 | XP_011513479.1 | 14 | 2015 | |||
XM_011515178.1 | XP_011513480.1 | 14 | 2226 | |||
XM_011515179.2 | XP_011513481.1 | 14 | 2006 | |||
XM_011515181.2 | XP_011513483.1 | 14 | 2006 | |||
XM_011515182.2 | XP_011513484.1 | 14 | 1801 | |||
XM_011515183.1 | XP_011513485.1 | 13 | 1895 | |||
XM_011515184.2 | XP_011513486.1 | 14 | 1801 | |||
XM_017011833.1 | XP_016867322.1 | 13 | 1727 | |||
XM_017011834.1 | XP_016867323.1 | 13 | 1918 | |||
XM_017011835.1 | XP_016867324.1 | 12 | 1811 | |||
GenBank mRNA | AK024482.1 | - | 11 | 3068 | ||
AK131083.1 | - | 10 | 4267 | |||
AL133088.1 | - | 6 | 641 | |||
BC007209.1 | - | 9 | 1199 | |||
BC015632.2 | - | 13 | 1718 | 3839985 | ||
BC023561.2 | - | 9 | 959 |