Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079653.1 | NP_001073121.1 | 8 | 1271 | ||
NM_152744.3 | NP_689957.3 | 34 | 5126 | |||
XM_011515188.1 | XP_011513490.1 | 28 | 3967 | |||
XM_011515189.1 | XP_011513491.1 | 28 | 3964 | |||
XM_011515190.1 | XP_011513492.1 | 28 | 3967 | |||
XM_017011837.1 | XP_016867326.1 | 28 | 3967 | |||
XM_017011838.1 | XP_016867327.1 | 28 | 3967 | |||
XM_017011839.1 | XP_016867328.1 | 28 | 3964 | |||
GenBank mRNA | AK074077.1 | - | 1 | 66 | ||
AL833354.1 | - | 8 | 1268 | |||
AY310398.1 | - | 34 | 5126 |