Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152864.3 | NP_690603.3 | 7 | 1344 | ||
XM_005260192.1 | XP_005260249.1 | 6 | 1221 | |||
XM_005260194.3 | XP_005260251.1 | 6 | 1680 | |||
XM_011528527.1 | XP_011526829.1 | 7 | 1552 | |||
XM_011528528.2 | XP_011526830.1 | 7 | 2014 | |||
XM_011528529.2 | XP_011526831.1 | 7 | 2000 | |||
XM_017027636.1 | XP_016883125.1 | 7 | 1522 | |||
XM_017027637.1 | XP_016883126.1 | 7 | 2004 | |||
XM_017027638.1 | XP_016883127.1 | 7 | 1764 | |||
XM_017027639.1 | XP_016883128.1 | 7 | 1750 | |||
GenBank mRNA | AK126246.1 | - | 7 | 1267 | ||
BC041812.1 | - | 7 | 1342 | 5302801 |