Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152888.2 | NP_690848.1 | 45, 46 | 3801 | ||
XM_011516883.2 | XP_011515185.1 | 44, 45 | 3819 | |||
XM_011516884.2 | XP_011515186.1 | 43, 44 | 3780 | |||
XM_011516885.2 | XP_011515187.1 | 43, 44 | 3765 | |||
XM_011516886.2 | XP_011515188.1 | 45, 46 | 3731 | |||
XM_011516887.2 | XP_011515189.1 | 41, 42 | 2699 | |||
XM_011516888.2 | XP_011515190.1 | 44, 45 | 3822 | |||
XM_011516889.2 | XP_011515191.1 | 34, 35 | 1940 | |||
XM_017013150.1 | XP_016868639.1 | 45, 46 | 3692 | |||
XM_017013151.1 | XP_016868640.1 | 41, 42 | 3657 | |||
XM_017013152.1 | XP_016868641.1 | 40, 41 | 2689 | |||
GenBank mRNA | AF406780.1 | - | 45, 46 | 3788 | ||
BC042075.1 | - | 19, 20 | 1026 | |||
BC144535.1 | - | 33, 34 | 2459 | 9053062 |