Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168271.1 | NP_001161743.1 | 4 | 646 | ||
NM_153002.2 | NP_694547.2 | 4 | 646 | |||
XM_005247165.2 | XP_005247222.1 | 5 | 923 | |||
XM_011512484.2 | XP_011510786.1 | 6 | 996 | |||
XM_011512487.1 | XP_011510789.1 | 4 | 548 | |||
XM_017005795.1 | XP_016861284.1 | 5 | 959 | |||
XM_017005796.1 | XP_016861285.1 | 5 | 968 | |||
XM_017005797.1 | XP_016861286.1 | 5 | 923 | |||
GenBank mRNA | AF488739.1 | - | 4 | 449 | ||
AY255565.1 | - | 1 | 38 | |||
BC113701.1 | - | 4 | 469 | 8322753 | ||
BC143606.1 | - | 4 | 469 | 9052119 |