Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168271.1 | NP_001161743.1 | 9 | 2345 | ||
NM_153002.2 | NP_694547.2 | 9 | 2357 | |||
XM_005247165.2 | XP_005247222.1 | 10 | 2634 | |||
XM_011512484.2 | XP_011510786.1 | 11 | 2707 | |||
XM_011512487.1 | XP_011510789.1 | 9 | 2259 | |||
XM_011512488.2 | XP_011510790.1 | 2 | 1326 | |||
XM_017005795.1 | XP_016861284.1 | 10 | 2670 | |||
XM_017005796.1 | XP_016861285.1 | 10 | 2679 | |||
XM_017005797.1 | XP_016861286.1 | 10 | 2622 | |||
GenBank mRNA | AB209362.1 | - | 4 | 1521 | ||
AF488739.1 | - | 9 | 2160 | |||
BC113701.1 | - | 9 | 2180 | 8322753 | ||
BC143606.1 | - | 9 | 2168 | 9052119 | ||
BX115336.1 | - | 1 | 106 |