Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166271.2 | NP_001159743.1 | 3 | 2306 | ||
NM_001286792.1 | NP_001273721.1 | 5 | 2630 | |||
NM_153023.3 | NP_694568.1 | 2 | 542 | |||
GenBank mRNA | AK055770.1 | - | 2 | 510 | ||
BC109290.1 | - | 1 | 252 | 40034357 | ||
BC109291.1 | - | 1 | 252 | 40034359 | ||
BX648244.1 | - | 5 | 2452 |