Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153256.3 | NP_694988.3 | 4 | 2994 | ||
XM_011519438.2 | XP_011517740.1 | 5 | 3313 | |||
XM_017016028.1 | XP_016871517.1 | 4 | 3980 | |||
GenBank mRNA | BC048800.1 | - | 4 | 2994 | 5215506 |