Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145160.1 | NP_001138632.1 | 3, 4 | 502 | ||
NM_003290.2 | NP_003281.1 | 2, 3 | 517 | |||
XM_005260042.3 | XP_005260099.1 | 2, 3 | 299 | |||
XM_006722865.2 | XP_006722928.1 | 3, 4 | 502 | |||
XM_011528231.2 | XP_011526533.1 | 3, 4 | 415 | |||
XM_011528232.1 | XP_011526534.1 | 2, 3 | 1735 | |||
XM_017027188.1 | XP_016882677.1 | 3, 4 | 533 | |||
XM_017027189.1 | XP_016882678.1 | 2, 3 | 2368 | |||
GenBank mRNA | AF186109.1 | - | 2, 3 | 279 | ||
AK023385.1 | - | 3, 4 | 471 | |||
AK301285.1 | - | 2, 3 | 347 | |||
AK307538.1 | - | 2, 3 | 281 | |||
AK315076.1 | - | 2, 3 | 321 | |||
BC002827.2 | - | 2, 3 | 324 | 3637300 | ||
BC037576.1 | - | 2, 3 | 392 | 5582453 | ||
BC067225.1 | - | 2, 3 | 346 | 4772340 | ||
BT019634.1 | - | 2, 3 | 276 | |||
DA032511.1 | - | 3, 4 | 502 | |||
FJ224305.1 | - | 2, 3 | 326 | |||
X05276.1 | - | 2, 3 | 326 |