Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153267.4 | NP_694999.3 | 3 | 954 | ||
NR_125850.1 | 3 | 954 | ||||
XM_011518471.1 | XP_011516773.1 | 3 | 1016 | |||
XM_011518475.2 | XP_011516777.1 | 3 | 1016 | |||
XM_017014587.1 | XP_016870076.1 | 3 | 1016 | |||
GenBank mRNA | AK304040.1 | - | 3 | 717 | ||
AK304531.1 | - | 3 | 542 | |||
BC016383.1 | - | 3 | 672 | 4696722 | ||
BC063634.1 | - | 3 | 931 | 4399866 |