Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153325.3 | NP_697020.2 | 2 | 390 | ||
GenBank mRNA | AF525927.1 | - | 2 | 352 | ||
AK311802.1 | - | 2 | 390 | |||
AY122477.1 | - | 2 | 340 | |||
BC128195.1 | - | 2 | 352 | 40113902 | ||
BC128196.1 | - | 2 | 352 | 40113904 |