Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORAOV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153451.2 | NP_703152.1 | 2 | 230 | ||
XM_006718470.2 | XP_006718533.1 | 2 | 187 | |||
GenBank mRNA | AF503940.1 | - | 2 | 180 | ||
AK026600.1 | - | 2 | 249 | |||
AK126490.1 | - | 1 | 85 | |||
AK294004.1 | - | 2 | 178 | |||
AK311918.1 | - | 2 | 180 | |||
BC006448.2 | - | 2 | 146 | |||
BC019024.1 | - | 2 | 147 | 4544931 | ||
BC065542.1 | - | 2 | 173 | 6101144 | ||
EF639695.1 | - | 2 | 80 |