Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193380.1 | NP_001180309.1 | 15 | 1586 | ||
NM_153480.1 | NP_705613.1 | 15 | 1513 | |||
NM_153481.1 | NP_705614.1 | 13 | 1377 | |||
NM_153483.2 | NP_705616.2 | 16 | 1532 | |||
NR_104198.1 | 14 | 1643 | ||||
XM_006712975.1 | XP_006713038.1 | 14 | 1339 | |||
XM_006712976.1 | XP_006713039.1 | 14 | 1477 | |||
XM_011533361.2 | XP_011531663.1 | 15 | 1429 | |||
XM_011533362.2 | XP_011531664.1 | 14 | 1293 | |||
GenBank mRNA | AF458069.1 | - | 15 | 1513 | ||
AK074238.1 | - | 15 | 1567 | |||
AK096793.1 | - | 14 | 1621 | |||
AK096801.1 | - | 13 | 1357 | |||
AK297697.1 | - | 15 | 1552 | |||
AK314476.1 | - | 15 | 1498 | |||
AY358451.1 | - | 15 | 1513 | |||
BC035478.1 | - | 16 | 1525 | |||
BC137077.1 | - | 16 | 1475 | 9021074 | ||
BC137078.1 | - | 16 | 1475 | 9021075 |