Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153703.4 | NP_714914.2 | 4 | 749 | ||
XM_005270457.3 | XP_005270514.1 | 5 | 778 | |||
XM_011540677.2 | XP_011538979.1 | 1 | 13 | |||
XM_017000294.1 | XP_016855783.1 | 4 | 1099 | |||
GenBank mRNA | AK296649.1 | - | 5 | 644 | ||
AK299375.1 | - | 6 | 834 | |||
AK314096.1 | - | 4 | 343 | |||
AY313607.1 | - | 4 | 588 | |||
AY358294.1 | - | 6 | 865 | |||
BC028292.1 | - | 4 | 572 | 4938380 | ||
BC030608.2 | - | 4 | 590 | 4826932 |