Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153703.4 | NP_714914.2 | 11 | 2895 | ||
XM_005270457.3 | XP_005270514.1 | 12 | 2924 | |||
XM_011540677.2 | XP_011538979.1 | 7 | 2049 | |||
XM_017000294.1 | XP_016855783.1 | 11 | 3245 | |||
GenBank mRNA | AK296649.1 | - | 12 | 2790 | ||
AY358294.1 | - | 13 | 3011 | |||
BC024024.1 | - | 1 | 647 | |||
BC028292.1 | - | 11 | 2718 | 4938380 | ||
BC030608.2 | - | 11 | 2736 | 4826932 |