Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003310.2 | NP_003301.1 | 4 | 500 | ||
XM_006711893.2 | XP_006711956.1 | 2 | 405 | |||
XM_011510383.2 | XP_011508685.1 | 5 | 636 | |||
XM_011510384.2 | XP_011508686.1 | 5 | 529 | |||
XM_011510385.2 | XP_011508687.1 | 4 | 529 | |||
XM_017004818.1 | XP_016860307.1 | 4 | 601 | |||
GenBank mRNA | AF019952.1 | - | 4 | 458 | ||
AK026522.1 | - | 4 | 441 | |||
AK126288.1 | - | 7 | 1489 | |||
AK222652.1 | - | 4 | 436 | |||
BC002485.2 | - | 4 | 408 | 3349430 | ||
DA349228.1 | - | 4 | 500 |