Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304376.2 | NP_001291305.1 | 6 | 603 | ||
NM_001318481.1 | NP_001305410.1 | 6 | 603 | |||
NM_153837.3 | NP_722579.1 | 6 | 1075 | |||
XM_011522949.2 | XP_011521251.1 | 6 | 619 | |||
XM_011522950.2 | XP_011521252.1 | 6 | 645 | |||
GenBank mRNA | AK128298.1 | - | 6 | 754 | ||
AK160368.1 | - | 6 | 585 | |||
AK172765.1 | - | 2 | 92 | |||
AK292783.1 | - | 6 | 595 | |||
AK301357.1 | - | 6 | 523 | |||
AK303661.1 | - | 6 | 595 | |||
AY140956.1 | - | 5 | 500 | |||
AY358447.1 | - | 6 | 582 | |||
BC032401.1 | - | 6 | 1023 | 5210813 |
Set Membership: |
G-protein coupled receptor |