Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017434.1 | NP_001017434.1 | 3 | 460 | ||
NM_170725.2 | NP_733843.1 | 3 | 1137 | |||
XM_005270333.1 | XP_005270390.1 | 4 | 1236 | |||
XM_011544159.2 | XP_011542461.1 | 3 | 1138 | |||
XM_011544161.2 | XP_011542463.1 | 3 | 2078 | |||
GenBank mRNA | AK123219.1 | - | 3 | 507 | ||
AK298248.1 | - | 4 | 1237 | |||
BC063785.1 | - | 3 | 1142 | 6173868 | ||
BX647065.1 | - | 3 | 449 |