Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172037.4 | NP_742034.1 | 2 | 1190 | ||
GenBank mRNA | AF456765.1 | - | 2 | 762 | ||
AY358270.1 | - | 2 | 582 | |||
BC067131.1 | - | 2 | 690 | 6153778 |