Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144766.2 | NP_001138238.1 | 2 | 153 | ||
NM_003315.3 | NP_003306.3 | 2 | 385 | |||
XM_011525167.2 | XP_011523469.1 | 3 | 315 | |||
XM_011525168.2 | XP_011523470.1 | 3 | 275 | |||
XM_011525169.2 | XP_011523471.1 | 2 | 127 | |||
XM_017024994.1 | XP_016880483.1 | 3 | 309 | |||
GenBank mRNA | AB209834.1 | - | 2 | 71 | ||
AK291047.1 | - | 2 | 86 | |||
AK296602.1 | - | 2 | 107 | |||
AK298860.1 | - | 2 | 101 | |||
AK308016.1 | - | 2 | 139 | |||
BC011837.2 | - | 2 | 104 | 3945721 | ||
BC033772.1 | - | 2 | 130 | 3924081 | ||
BP378441.1 | - | 2 | 134 | |||
BX647209.1 | - | 2 | 341 | |||
DB453150.1 | - | 2 | 385 | |||
U46571.1 | - | 2 | 100 |