Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172193.2 | NP_751943.1 | 7 | 722 | ||
XM_011536422.2 | XP_011534724.1 | 7 | 645 | |||
GenBank mRNA | AF111806.1 | - | 7 | 660 | ||
AK127202.1 | - | 7 | 645 | |||
AL833437.1 | - | 2 | 152 | |||
BC031270.1 | - | 7 | 685 | |||
BC101595.1 | - | 7 | 722 | 8069101 | ||
BC101597.1 | - | 7 | 722 | 8069103 |