Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161812.1 | NP_001155284.1 | 4 | 608 | ||
NM_001161814.1 | NP_001155286.1 | 5 | 693 | |||
NM_172370.4 | NP_758958.3 | 4 | 453 | |||
XM_005254042.1 | XP_005254099.1 | 5 | 745 | |||
GenBank mRNA | AY138546.1 | - | 4 | 426 | ||
AY138547.1 | - | 5 | 693 | |||
AY170469.2 | - | 5 | 515 | |||
AY170470.2 | - | 6 | 609 | |||
AY170471.1 | - | 6 | 691 | |||
AY223901.1 | - | 5 | 745 | |||
BC121091.1 | - | 4 | 421 | 40119921 | ||
DQ343761.1 | - | 4 | 700 | |||
DQ357223.1 | - | 4 | 445 | |||
DQ386869.1 | - | 4 | 508 | |||
DQ386870.1 | - | 4 | 554 |